Product Details
- SNP ID
-
rs5020842
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.15:75029969 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCCATCCCACGCAGCCAGCGTTTGG[C/T]GGGAGGGGGGTTGTGAGGAAAGGGT
- Phenotype
-
MIM: 609854
MIM: 613766
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
PPCDC
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs77489018] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- PPCDC
- Gene Name
- phosphopantothenoylcysteine decarboxylase
- Gene
- SCAMP5
- Gene Name
- secretory carrier membrane protein 5
There are no transcripts associated with this gene.
View Full Product Details