Product Details

SNP ID
rs34130645
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:84889002 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAGGAATAAAGGCGCTTTAGCCAA[C/T]ACGCACAGGCACATGAGCCTCGAGG
Phenotype
MIM: 606207
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SLC28A1 PubMed Links

Gene Details

Gene
SLC28A1
Gene Name
solute carrier family 28 member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001287761.1 Intron NP_001274690.1
NM_001287762.1 Intron NP_001274691.1
NM_001321721.1 Intron NP_001308650.1
NM_001321722.1 Intron NP_001308651.1
NM_004213.4 Intron NP_004204.3
NM_201651.2 Intron NP_964014.1
XM_011522203.2 Intron XP_011520505.1
XM_011522204.2 Intron XP_011520506.1
XM_011522205.2 Intron XP_011520507.1
XM_011522206.2 Intron XP_011520508.1
XM_011522207.2 Intron XP_011520509.1
XM_011522208.2 Intron XP_011520510.1
XM_011522209.2 Intron XP_011520511.1
XM_011522210.2 Intron XP_011520512.1
XM_011522211.2 Intron XP_011520513.1
XM_011522212.1 Intron XP_011520514.1
XM_011522214.2 Intron XP_011520516.1
XM_011522215.2 Intron XP_011520517.1
XM_011522216.2 Intron XP_011520518.1
XM_011522217.1 Intron XP_011520519.1
XM_011522218.2 Intron XP_011520520.1
XM_017022723.1 Intron XP_016878212.1

View Full Product Details