Product Details
- SNP ID
-
rs9505892
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:169217016 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- AGGCAGGAGCCTCCCCGACGCCACA[A/G]CGGCACAAGCAGCAGCTAAAGCACC
- Phenotype
-
MIM: 188061
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
LOC101929523
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs114924628] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- LOC101929523
- Gene Name
- uncharacterized LOC101929523
There are no transcripts associated with this gene.
- Gene
- THBS2
- Gene Name
- thrombospondin 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_003247.3 |
4645 |
UTR 3 |
|
|
NP_003238.2 |
View Full Product Details