Product Details
- SNP ID
-
hCV317452
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:1149177 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AAAAATTAAAAACATTTTTTCATTA[A/G]AACACCTGATCTGCAGATTCACAGT
- Phenotype
-
MIM: 610699
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ZFAND2A
PubMed Links
Gene Details
- Gene
- ZFAND2A
- Gene Name
- zinc finger AN1-type containing 2A
There are no transcripts associated with this gene.
View Full Product Details