Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_139159.4 | 1229 | Intron | NP_631898.3 | ||
XM_005259673.2 | 1229 | Intron | XP_005259730.1 | ||
XM_011528405.1 | 1229 | Intron | XP_011526707.1 | ||
XM_011528406.1 | 1229 | Intron | XP_011526708.1 | ||
XM_011528408.1 | 1229 | Intron | XP_011526710.1 | ||
XM_011528410.1 | 1229 | Intron | XP_011526712.1 | ||
XM_017027453.1 | 1229 | Intron | XP_016882942.1 | ||
XM_017027454.1 | 1229 | Intron | XP_016882943.1 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001242901.1 | 1229 | UTR 3 | NP_001229830.1 |