Product Details
- SNP ID
-
rs11912172
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.22:45674928 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GAGTAATAGAAGTGTCATGCAGTGG[G/T]AAGAGCAGTGGCTTTGGAACCATGG
- Phenotype
-
MIM: 611150
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
ATXN10
PubMed Links
Gene Details
- Gene
- ATXN10
- Gene Name
- ataxin 10
View Full Product Details