Product Details

SNP ID
rs8185
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.14:74942086 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
TAGGGGGGGCAGGGTGGTGGCACCT[C/T]GGCCGGAAAGAACAATGTCCCAGTA
Phenotype
MIM: 601121
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PGF PubMed Links

Gene Details

Gene
PGF
Gene Name
placental growth factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001207012.1 1655 UTR 3 NP_001193941.1
NM_001293643.1 1655 UTR 3 NP_001280572.1
NM_002632.5 1655 UTR 3 NP_002623.2

View Full Product Details