Product Details
- SNP ID
-
rs3110824
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:135673520 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTATGTCACATTTATTGTTTATATG[C/T]ACAACCTTTACTTGGCACCAGATCG
- Phenotype
-
MIM: 604309
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
C7orf73
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs79580568] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- C7orf73
- Gene Name
- chromosome 7 open reading frame 73
- Gene
- SLC13A4
- Gene Name
- solute carrier family 13 member 4
There are no transcripts associated with this gene.
View Full Product Details