Product Details

SNP ID
rs538319
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:6091604 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTGATTATATTGATTCTCATTCTCA[A/C]CCCCATATTAATTTTATTCTTGTTT
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
USP42 PubMed Links

Gene Details

Gene
USP42
Gene Name
ubiquitin specific peptidase 42
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032172.2 Intron NP_115548.1
XM_005249883.4 Intron XP_005249940.1
XM_006715790.1 Intron XP_006715853.1
XM_006715791.2 Intron XP_006715854.1
XM_011515573.1 Intron XP_011513875.1
XM_011515574.1 Intron XP_011513876.1
XM_011515577.1 Intron XP_011513879.1
XM_011515578.1 Intron XP_011513880.1

View Full Product Details