Product Details
- SNP ID
-
rs446791
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:151273365 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CTTCACATCCTTCAGTAGTTACCTG[C/T]TGGATAACCTATAACCTGCAGAAGT
- Phenotype
-
MIM: 603525
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
NMI
PubMed Links
Gene Details
- Gene
- NMI
- Gene Name
- N-myc and STAT interactor
View Full Product Details