Product Details
- SNP ID
-
rs355467
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:67486007 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- AAAGCCCCCCGAAATGTTTCAAAGG[C/T]ATTTTTATAACTTAATAAGACAAAA
- Phenotype
-
MIM: 117141
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CENPC
PubMed Links
Gene Details
- Gene
- CENPC
- Gene Name
- centromere protein C
View Full Product Details