Product Details

SNP ID
rs654332
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:13317815 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAAGAGAGACGATGTTAGCAGCAG[A/C]GCCAGGGAAAGCGGCGCACTCTTCA
Phenotype
MIM: 607703
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
NUP210 PubMed Links

Gene Details

Gene
NUP210
Gene Name
nucleoporin 210
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024923.3 Intron NP_079199.2
XM_005264994.3 Intron XP_005265051.1
XM_011533538.2 Intron XP_011531840.1
XM_017006018.1 Intron XP_016861507.1
XM_017006019.1 Intron XP_016861508.1
XM_017006020.1 Intron XP_016861509.1
XM_017006021.1 Intron XP_016861510.1

View Full Product Details