Product Details

SNP ID
rs510304
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:38455439 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGTCCCCCGCCGCCCTCGGTGGCA[C/G]CCGCCTGGAGTCTGGGAAACCCTCG
Phenotype
MIM: 602730 MIM: 604049
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ACVR2B PubMed Links

Gene Details

Gene
ACVR2B
Gene Name
activin A receptor type 2B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001106.3 Intron NP_001097.2
XM_005265583.3 Intron XP_005265640.1
XM_017007514.1 Intron XP_016863003.1
XM_017007515.1 Intron XP_016863004.1
XM_017007516.1 Intron XP_016863005.1
Gene
ACVR2B-AS1
Gene Name
ACVR2B antisense RNA 1
There are no transcripts associated with this gene.

Gene
XYLB
Gene Name
xylulokinase
There are no transcripts associated with this gene.

View Full Product Details