Product Details

SNP ID
rs1078109
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:57592583 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTCAGTGGAAGGCACTTGGTATGGG[A/G]AAGGCTGATGGAGGTAGGGCTGCCT
Phenotype
MIM: 602821
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KIF5A PubMed Links

Gene Details

Gene
KIF5A
Gene Name
kinesin family member 5A
There are no transcripts associated with this gene.

Gene
PIP4K2C
Gene Name
phosphatidylinositol-5-phosphate 4-kinase type 2 gamma
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001146258.1 Intron NP_001139730.1
NM_001146259.1 Intron NP_001139731.1
NM_001146260.1 Intron NP_001139732.1
NM_024779.4 Intron NP_079055.3
XM_005269152.2 Intron XP_005269209.1
XM_011538747.1 Intron XP_011537049.1
XM_017019973.1 Intron XP_016875462.1

View Full Product Details