Product Details
- SNP ID
-
rs725687
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.9:37755887 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TATACTGTGAACTCTTAAGTGTTTA[A/G]GTAAATGTAGGTATAAGCCCTCCTA
- Phenotype
-
MIM: 616919
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
FRMPD1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs140500414] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- FRMPD1
- Gene Name
- FERM and PDZ domain containing 1
There are no transcripts associated with this gene.
- Gene
- TRMT10B
- Gene Name
- tRNA methyltransferase 10B
View Full Product Details