Product Details

SNP ID
rs764190
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:38760346 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCATTTCCCTTTGGGTCTGGGTCTA[A/G]GCCGGGGCCTTGTCTGAATAGGCTT
Phenotype
MIM: 603261 MIM: 602176
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC100287808 PubMed Links

Gene Details

Gene
LOC100287808
Gene Name
uncharacterized LOC100287808
There are no transcripts associated with this gene.

Gene
PIP4K2B
Gene Name
phosphatidylinositol-5-phosphate 4-kinase type 2 beta
There are no transcripts associated with this gene.

Gene
PSMB3
Gene Name
proteasome subunit beta 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002795.3 Intron NP_002786.2

View Full Product Details