Product Details
- SNP ID
-
rs176090
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.18:24143168 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TACCCTATGGCCTCAAGACTCTGCT[A/C]GAGGGAATTAGCAGAGCTGTTCTCA
- Phenotype
-
MIM: 612135
- Polymorphism
- A/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
CABYR
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs76208354] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CABYR
- Gene Name
- calcium binding tyrosine phosphorylation regulated
- Gene
- TTC39C
- Gene Name
- tetratricopeptide repeat domain 39C
There are no transcripts associated with this gene.
View Full Product Details