Product Details
- SNP ID
-
rs104893760
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:87260022 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ACTACTTCTTTCTCCAGATTCAGTT[A/C]TTCAGCCATCCTCATGATCTCTTGA
- Phenotype
-
MIM: 609512
MIM: 173110
- Polymorphism
- A/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
CHMP2B
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs76296626] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CHMP2B
- Gene Name
- charged multivesicular body protein 2B
There are no transcripts associated with this gene.
- Gene
- POU1F1
- Gene Name
- POU class 1 homeobox 1
View Full Product Details