Product Details

SNP ID
rs4916497
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.3:196471662 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
TACCGCTGAGCTGTGCAGAAGCTCA[C/T]GTGTCTATGCAGTCCTTACAATCAC
Phenotype
MIM: 612688
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RNF168 PubMed Links

Gene Details

Gene
RNF168
Gene Name
ring finger protein 168
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152617.3 2468 UTR 3 NP_689830.2

View Full Product Details