Product Details
- SNP ID
-
rs35880938
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
28
- Location
-
Chr.1:42534959 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GATAGAAGAGAAACCAGTACAAAAG[A/G]TCAAGAGACAAAGTACAAGATTTGA
- Phenotype
-
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
CCDC30
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs200019270] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CCDC30
- Gene Name
- coiled-coil domain containing 30
View Full Product Details