Product Details

SNP ID
rs2319797
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:68228789 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GACATAAATCCAGCACATAACATTG[A/T]ATCAGTCACAAAGCCAGAGTATGCA
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
TMPRSS11B PubMed Links

Gene Details

Gene
TMPRSS11B
Gene Name
transmembrane protease, serine 11B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_182502.3 1178 Missense Mutation ACA,TCA T348S NP_872308.2
XM_011531608.2 1178 Missense Mutation ACA,TCA T348S XP_011529910.1
XM_011531609.1 1178 Intron XP_011529911.1

View Full Product Details