Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
dhhc-1
Entrez Gene ID
181606
Gene Name
Palmitoyltransferase
Gene Aliases
CELE_F09B12.2
Chromosome Location
Chr.X: 15098066 - 15100423 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_078109.6 NP_510510.1 1 - 2 105 67

View Full Product Details