Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
dhs-30
Entrez Gene ID
181761
Gene Name
DeHydrogenases, Short chain
Gene Aliases
CELE_T25G12.7
Chromosome Location
Chr.X: 17230702 - 17235838 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_078392.6 NP_510793.2 2 - 3 171 100

View Full Product Details