Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
pnk-1
Entrez Gene ID
172327
Gene Name
protein-coding
Gene Aliases
CELE_C10G11.5
Chromosome Location
Chr.I: 6295063 - 6297636 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001380502.1 NP_001367496.1 8 - 9 1281 82
NM_170878.5 NP_740867.1 8 - 9 1268 82
NM_170879.8 NP_740868.1 8 - 9 1273 82

View Full Product Details