Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- stim-1
- Entrez Gene ID
-
175201
- Gene Name
- protein-coding
- Gene Aliases
-
CELE_Y55B1BM.1
- Chromosome Location
-
Chr.III: 408094 - 418749 on Build WBcel235
- Gene Symbol
- gei-4
- Entrez Gene ID
-
175195
- Gene Name
- protein-coding
- Gene Aliases
-
CELE_W07B3.2
- Chromosome Location
-
Chr.III: 344306 - 418744 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_001384004.1 |
|
1 - 2 |
131 |
137 |
View Full Product Details