Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
pqn-25
Entrez Gene ID
175759
Gene Name
Prion-like-(Q/N-rich) domain-bearing protein 25
Gene Aliases
CELE_D1044.3
Chromosome Location
Chr.III: 5509170 - 5512602 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_171123.6 NP_741140.1 9 - 10 1697 84

View Full Product Details