Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
hipr-1
Entrez Gene ID
176224
Gene Name
protein-coding
Gene Aliases
CELE_ZK370.3
Chromosome Location
Chr.III: 8720201 - 8725347 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_066524.6 NP_498925.1 6 - 7 1624 94
NM_171215.7 NP_741253.1 6 - 7 1625 94

View Full Product Details