Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
hil-2
Entrez Gene ID
177390
Gene Name
protein-coding
Gene Aliases
CELE_Y73B6BL.9
Chromosome Location
Chr.IV: 6294240 - 6301791 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001383102.1 1 - 2 267 76
GenBank mRNA AF017811.1 - 1 - 2 246 76

View Full Product Details