Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srh-51
Entrez Gene ID
191862
Gene Name
Serpentine Receptor, class H
Gene Aliases
CELE_C10G11.3
Chromosome Location
Chr.I: 6300493 - 6302298 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_059416.2 NP_491817.1 1 - 2 222 128

View Full Product Details