Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
pqn-21
Entrez Gene ID
172398
Gene Name
protein-coding
Gene Aliases
CELE_C37A2.5
Chromosome Location
Chr.I: 6773973 - 6780151 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001025858.4 NP_001021029.1 11 - 12 2390 148
NM_001306351.3 NP_001293280.1 9 - 10 1728 148
NM_001306352.3 NP_001293281.1 7 - 8 1281 148
NM_059539.6 NP_491940.2 10 - 11 2203 148
GenBank mRNA AF170425.1 - 11 - 12 2159 148

View Full Product Details