Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
pqn-21
Entrez Gene ID
172398
Gene Name
protein-coding
Gene Aliases
CELE_C37A2.5
Chromosome Location
Chr.I: 6773973 - 6780151 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001025858.4 NP_001021029.1 2 - 3 597 94
NM_059539.6 NP_491940.2 2 - 3 356 94
GenBank mRNA AF170425.1 - 2 - 3 366 94

View Full Product Details