Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
pqn-21
Entrez Gene ID
172398
Gene Name
protein-coding
Gene Aliases
CELE_C37A2.5
Chromosome Location
Chr.I: 6773973 - 6780151 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001025858.4 NP_001021029.1 5 - 6 1337 151
NM_001306351.3 NP_001293280.1 3 - 4 675 151
NM_001306352.3 NP_001293281.1 1 - 2 228 151
NM_059539.6 NP_491940.2 5 - 6 1096 151
GenBank mRNA AF170425.1 - 5 - 6 1106 151

View Full Product Details