Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
hint-1
Entrez Gene ID
184760
Gene Name
Histidine triad nucleotide-binding protein 1
Gene Aliases
CELE_F21C3.3
Chromosome Location
Chr.I: 7279666 - 7280489 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_059655.6 NP_492056.1 2 - 3 258 110

View Full Product Details