Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
lrp-1
Entrez Gene ID
172520
Gene Name
Low-density lipoprotein receptor-related protein
Gene Aliases
CELE_F29D11.1
Chromosome Location
Chr.I: 7614176 - 7634142 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001383375.1 21 - 22 9765 109

View Full Product Details