Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
sin-3
Entrez Gene ID
172628
Gene Name
protein-coding
Gene Aliases
CELE_F02E9.4
Chromosome Location
Chr.I: 8416816 - 8425051 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001128970.4 NP_001122442.1 15 - 16 3558 79
NM_059883.8 NP_492284.2 15 - 16 3553 79

View Full Product Details