Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ces-1
Entrez Gene ID
185718
Gene Name
CEll death Specification
Gene Aliases
CELE_F43G9.11
Chromosome Location
Chr.I: 8633897 - 8636169 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_059937.3 NP_492338.1 1 - 2 122 98

View Full Product Details