Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
twk-30
Entrez Gene ID
185335
Gene Name
TWiK family of potassium channels
Gene Aliases
CELE_F36A2.4
Chromosome Location
Chr.I: 8808950 - 8811647 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_059980.6 NP_492381.3 5 - 6 789 110

View Full Product Details