Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
nduf-5
Entrez Gene ID
266824
Gene Name
NADH Ubiquinone oxidoreductase Fe-S protein
Gene Aliases
CELE_Y54E10BL.5
Chromosome Location
Chr.I: 2996631 - 3003512 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_058684.7 NP_491085.1 2 - 3 337 77

View Full Product Details