Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
arx-7
Entrez Gene ID
171882
Gene Name
Actin-related protein 2/3 complex subunit 5
Gene Aliases
CELE_M01B12.3
Chromosome Location
Chr.I: 3069322 - 3078677 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001381571.2 NP_001367661.1 7 - 8 1398 63

View Full Product Details