Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
pqn-72
Entrez Gene ID
189086
Gene Name
protein-coding
Gene Aliases
CELE_W01B11.5
Chromosome Location
Chr.I: 3269402 - 3280883 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001129055.4 NP_001122527.1 6 - 7 867 147
NM_001313513.3 NP_001300442.1 5 - 6 352 147
NM_001313514.3 NP_001300443.1 5 - 6 808 147
NM_058736.5 NP_491137.2 5 - 6 349 147

View Full Product Details