Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
exoc-7
Entrez Gene ID
172023
Gene Name
Exocyst complex component 7
Gene Aliases
CELE_C43E11.8
Chromosome Location
Chr.I: 4242821 - 4247582 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_058936.5 NP_491337.1 3 - 4 789 100

View Full Product Details