Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
phat-2
Entrez Gene ID
172130
Gene Name
PHAryngeal gland Toxin-related
Gene Aliases
CELE_C46H11.9
Chromosome Location
Chr.I: 5046394 - 5048018 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_059108.6 NP_491509.1 1 - 2 113 105

View Full Product Details