Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
unc-26
Entrez Gene ID
178284
Gene Name
protein-coding
Gene Aliases
CELE_JC8.10
Chromosome Location
Chr.IV: 13261537 - 13272516 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001028094.4 NP_001023265.1 5 - 6 1501 84
GenBank mRNA AF283323.1 - 5 - 6 1523 84

View Full Product Details