Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
mec-2
Entrez Gene ID
180826
Gene Name
protein-coding
Gene Aliases
CELE_F14D12.4
Chromosome Location
Chr.X: 5567754 - 5590830 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001368504.4 NP_001355460.1 5 - 5 1029 60
NM_001368505.3 NP_001355457.1 9 - 10 1098 60
NM_001368511.3 NP_001355478.1 9 - 10 1074 60
NM_001368515.4 NP_001355479.1 9 - 9 1050 60
NM_001380971.2 NP_001368710.1 9 - 10 1215 60
NM_001380972.1 NP_001368712.1 10 - 11 1275 60
NM_001380974.1 NP_001368713.1 9 - 10 1074 60
NM_001380976.1 NP_001368714.1 9 - 10 1050 60
NM_171691.6 NP_741797.1 10 - 11 1318 60
GenBank mRNA U26735.1 - 10 - 11 1312 60

View Full Product Details