Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
mec-2
Entrez Gene ID
180826
Gene Name
protein-coding
Gene Aliases
CELE_F14D12.4
Chromosome Location
Chr.X: 5567754 - 5590830 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_171691.6 NP_741797.1 12 - 13 1470 107
GenBank mRNA U26735.1 - 12 - 13 1464 107

View Full Product Details