Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
lgc-47
Entrez Gene ID
181193
Gene Name
protein-coding
Gene Aliases
CELE_F47A4.1
Chromosome Location
Chr.X: 9801078 - 9807663 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_171756.4 NP_741878.1 4 - 5 383 63
NM_171757.7 NP_741879.1 4 - 5 602 63

View Full Product Details