Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
tag-10
Entrez Gene ID
175011
Gene Name
protein-coding
Gene Aliases
CELE_C31C9.1
Chromosome Location
Chr.II: 13632584 - 13636998 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001322756.4 NP_001309535.1 4 - 4 679 69
NM_001322757.3 NP_001309536.1 3 - 4 620 69
NM_064466.6 NP_496867.2 3 - 4 624 69
NM_182157.5 NP_871957.1 3 - 4 601 69

View Full Product Details