Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
clec-92
Entrez Gene ID
190182
Gene Name
C-type lectin domain-containing protein
Gene Aliases
CELE_Y52B11A.5
Chromosome Location
Chr.I: 11015483 - 11017537 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_060456.2 NP_492857.1 1 - 2 186 88

View Full Product Details