Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
abt-5
Entrez Gene ID
173081
Gene Name
ABC Transporter family
Gene Aliases
CELE_Y53C10A.9
Chromosome Location
Chr.I: 12011195 - 12018684 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001377649.1 NP_001364572.1 2 - 3 359 101

View Full Product Details