The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001136318.5 | NP_001129790.1 | 2 - 3 | 270 | 68 |
NM_060942.5 | NP_493343.1 | 4 - 5 | 575 | 68 |